1,2-Dibromoethane, 99%
1,2-Dibromoethane, 99%
1,2-Dibromoethane, 99%
1,2-Dibromoethane, 99%
Thermo Scientific Chemicals

1,2-Dibromoethane, 99%

CAS: 106-93-4 | C2H4Br2 | 187.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
10 L
Packaging:
View all
Plastic Drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 112790100
Price (USD)
-
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS106-93-4
IUPAC Name1,2-dibromoethane
Molecular FormulaC2H4Br2
InChI KeyPAAZPARNPHGIKF-UHFFFAOYSA-N
SMILESBrCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5375 to 1.5395 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only