본 제품은 LMO 제품으로, 고객 분께서 LMO 신고 시스템을 통해 직접 수입 신고를 진행해주셔야 합니다. 자세히보기
Product Image
Invitrogen™

MaV203 Competent Yeast Cells, Library Scale

MaV203 Competent Yeast Cells are designed for use with the ProQuest™ Two-Hybrid System (Figure 1). S. cerevisiae strain MaV203 contains자세히 알아보기
Have Questions?
카탈로그 번호수량
11281011
11281-011으로도 사용됨
2 x 0.55 mL
카탈로그 번호 11281011
11281-011으로도 사용됨
제품 가격(KRW)
-
수량:
2 x 0.55 mL
MaV203 Competent Yeast Cells are designed for use with the ProQuest™ Two-Hybrid System (Figure 1). S. cerevisiae strain MaV203 contains deletions in the endogenous GAL4 and GAL80 genes for use with GAL4-based two-hybrid systems. The strain also has leu2 and trp1 mutations for selection of ProQuest™ bait and prey vectors. MaV203 contains three GAL4 inducible reporter genes, providing four phenotypes, for easier identification of true interactors (Figure 2). The MaV203 competent cells feature:

• Transformation (250 μl of cells) efficiency yields >1 x 106 colonies
• Control DNA pMAB12 to confirm transformation efficiency and the preparation of selection media
• Pre-qualified PEG/Lithium Acetate to maximize number of trans-formants
• Competent cells formatted with carrier DNA already included
For Research Use Only. Not for use in diagnostic procedures.
사양
박테리아 또는 효모 균주MaV203
블루/화이트 스크리닝No
형식Tube(s)
고처리량 호환성Not High-throughput Compatible (Manual)
플라스미드 품질 개선No
제품 유형Yeast Cells
수량2 x 0.55 mL
배송 조건Dry Ice
종S. cerevisiae
Unit SizeEach
구성 및 보관
Store in ultra-cold freezer (-68 to -85°C).

자주 묻는 질문(FAQ)

Does the ProQuest Two-Hybrid System yeast strain, MaV203 have slow growth compared to other commonly used laboratory Saccharomyces cerevisiae strains?

To avoid interference of endogenous GAL4 and GAL80 proteins, MaV203 must carry deletions of the GAL4 and GAL80 genes. As a result of deletion of these two genes, MaV203 cells grow more slowly compared to yeast strains containing the wild-type version of these genes. Please note that the growth rate will depend on the type of growth medium used (e.g., YPD versus SC drop-out media).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Are the MaV203 yeast cells that come with the ProQuest Two-Hybrid System ready for transformation?

No, the MaV203 cells are provided as a glycerol stock. Cells from this stock need to be streaked on a YPD plate for growth at 30 degrees C. Resulting colonies can be used to prepare competent cells for transformation as per the ProQuest Two-Hybrid System manual. Alternatively, ready-to-use MaV203 Competent Yeast Cells, Subcloning Scale (Cat. No. 11445-012) or Library Scale (Cat. No. 11281-011) are available for purchase from Thermo Fisher Scientific.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I sequenced one of your vectors after PCR amplification and observed a difference from what is provided online (or in the manual). Should I be concerned?

Our vectors have not been completely sequenced. Your sequence data may differ when compared to what is provided. Known mutations that do not affect the function of the vector are annotated in public databases.

Are your vectors routinely sequenced?

No, our vectors are not routinely sequenced. Quality control and release criteria utilize other methods.

How was the reference sequence for your vectors created?

Sequences provided for our vectors have been compiled from information in sequence databases, published sequences, and other sources.