Dibromomethane, 99%
Dibromomethane, 99%
Dibromomethane, 99%
Acros Organics

Dibromomethane, 99%

CAS: 74-95-3 | CH2Br2 | 173.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
View all
Glass bottle
Catalog number 112831000
HSN 29033990
Price (INR)
4,823.00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS74-95-3
IUPAC Namedibromomethane
Molecular FormulaCH2Br2
InChI KeyFJBFPHVGVWTDIP-UHFFFAOYSA-N
SMILESBrCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5400 to 1.5420 (20°C, 589 nm)
Appearance (Color)Clear colorless to light brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only