Interested in a bulk or custom version of this product? Request pricing.

ADSL Recombinant Human Protein, N-His tag

Catalog number: 11287H07E50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Adenylosuccinate Lyase (ADSL), recombinant human protein is supplied in frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human ADSL (P30566-1) (Met 1-Leu 484) fused to a polyhistidine tag at the N-terminus.

Activity: The activity of this protein has not been tested.

Reconstitution: No reconstitution is needed. Store aliquots under sterile conditions at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles.

Formulation: PBS, 10% Glycerol, pH 7.4.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Other Proteins
Research Category: Other Research
Expression: E. coli
Form: Frozen
Gene Aliases: RP5-1042K10.8, AMPS, ASASE, ASL
Gene ID: 158
Gene Symbol: ADSL
Molecular Weight: 57 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Form: Recombinant
Protein Name: Adenylosuccinate Lyase
Protein Tag or Fusion: His Tag
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 1 vials of protein in suspension buffer. Store at -80 °C.

Documents

Manuals & protocols