2,6-Dibromopyridine, 98%
2,6-Dibromopyridine, 98%
2,6-Dibromopyridine, 98%
Thermo Scientific Chemicals

2,6-Dibromopyridine, 98%

CAS: 626-05-1 | C5H3Br2N | 236.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 112911000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS626-05-1
IUPAC Name2,6-dibromopyridine
Molecular FormulaC5H3Br2N
InChI KeyFEYDZHNIIMENOB-UHFFFAOYSA-N
SMILESBrC1=CC=CC(Br)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point117°C to 121°C
GC>=97.5 %
Appearance (Color)White to grey to yellow-brown
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only