2,6-Dibromopyridine, 98%
2,6-Dibromopyridine, 98%
2,6-Dibromopyridine, 98%
Thermo Scientific Chemicals

2,6-Dibromopyridine, 98%

CAS: 626-05-1 | C5H3Br2N | 236.89 g/mol
数量:
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 112915000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS626-05-1
IUPAC Name2,6-dibromopyridine
Molecular FormulaC5H3Br2N
InChI KeyFEYDZHNIIMENOB-UHFFFAOYSA-N
SMILESBrC1=CC=CC(Br)=N1
さらに表示
Infrared spectrumConforms
Melting point117°C to 121°C
GC>=97.5 %
Appearance (Color)White to grey to yellow-brown
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only