QSP Specialty Pipette Tips
Thermo Scientific™

QSP Specialty Pipette Tips

Specialty pipette tips designed for specific applications making it easier to concentrate on what is important—your research.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionVolume (Metric)Tip StyleSterilityUnit Size
112NXLG-QQSP 1000 Genomic Wide Orifice, Non-Filtered1000 μLGenomic/Wide OrificeNon-sterileCase of 10000
103-QQSP 10 Gel, Non-Filtered10 μLGel LoadingNon-sterileCase of 10000
118-N-QQSP 200 Genomic/Wide Orifice, Non-Filtered200 μLGenomic/Wide OrificeNon-sterileCase of 10000
118-N-QPK
also known as 118-N-Q
QSP 200 Genomic/Wide Orifice, Non-Filtered200 μLGenomic/Wide OrificeNon-sterilePack of 1000
T118R-QQSP 200 Genomic, Non-Filtered200 μLGenomic/Wide OrificeNon-sterileCase of 4800
T118R-QPK
also known as T118R-Q
QSP 200 Genomic, Non-Filtered200 μLGenomic/Wide OrificeNon-sterilePack of 960
T118RS-QQSP 200 Genomic, Non-Filtered200 μLGenomic/Wide OrificeSterileCase of 4800
T118RS-QPK
also known as T118RS-Q
QSP 200 Genomic, Non-Filtered200 μLGenomic/Wide OrificeSterilePack of 960
TF118-200-QQSP 200 Genomic, Filtered200 μLGenomic/Wide OrificeSterileCase of 4800
TF118-200-QPK
also known as TF118-200-Q
QSP 200 Genomic, Filtered200 μLGenomic/Wide OrificeSterilePack of 960
112NXLG-QPK
also known as 112NXLG-Q
QSP 1000 Genomic Wide Orifice, Non-Filtered1000 μLGenomic/Wide OrificeNon-sterilePack of 1000
T112NXLGR-QQSP 1000 Genomic, Non-Filtered1000 μLGenomic/Wide OrificeNon-sterileCase of 4800
T112NXLGR-QPK
also known as T112NXLGR-Q
QSP 1000 Genomic, Non-Filtered1000 μLGenomic/Wide OrificeNon-sterilePack of 960
T112NXLGRS-QQSP 1000 Genomic, Non-Filtered1000 μLGenomic/Wide OrificeSterileCase of 4800
T112NXLGRS-QPK
also known as T112NXLGRS-Q
QSP 1000 Genomic, Non-Filtered1000 μLGenomic/Wide OrificeSterilePack of 960
TF112G-1000-QQSP 1000 Genomic, Filtered1000 μLGenomic/Wide OrificeSterileCase of 4800
TF112G-1000-QPK
also known as TF112G-1000-Q
QSP 1000 Genomic, Filtered1000 μLGenomic/Wide OrificeSterilePack of 960
Catalog number 112NXLG-Q
Price (HKD)
-
Description:
QSP 1000 Genomic Wide Orifice, Non-Filtered
Volume (Metric):
1000 μL
Tip Style:
Genomic/Wide Orifice
Sterility:
Non-sterile
Unit Size:
Case of 10000
Thermo Scientific™ QSP specialty pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Specialty tips are crafted for specific applications and are designed to make your work more efficient and reproducible.

Gel Loading Specialty Tips
Loading acrylamide and agarose gels with standard pipette tips is a lengthy and tedious process. Avoid damaging your gel by choosing a specialty tip that offers a longer taper with a thinner distal end that is designed for these applications.

Genomic Wide Orifice Specialty Tips:
A problem posed when working with viscous liquids like tween or thick cell lysates is pipetting reliability due to the inability to draw and dispense fluid in a uniform manner. Our large, wide orifice tips are much wider than standard tips improving capability to draw and dispense in a consistent and uniform manner.

Extended Length Specialty Tips:
Extended length tips help prevent contamination by keeping the pipette shaft from entering the sample vessel. Extended length tips can provide laboratory solutions for dispensing samples into hard to reach vessels were a standard tip just cant reach.

Clean Claims:
QSP Pipette Tips are free of:

  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP

To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.

Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.

Specifications
Clean ClaimsDNase, RNase, DNA, and Pyrogen Free
DescriptionQSP 1000 Genomic Wide Orifice, Non-Filtered
Length (Metric)8.9cm
ColorNatural
FilteredNon-filtered
For Use With (Equipment)Universal Fit
Product LineQSP
SterilityNon-sterile
Tip StyleGenomic/Wide Orifice
Volume (Metric)1000 μL
Unit SizeCase of 10000