Interested in a bulk or custom version of this product? Request pricing.

SEMA5A Recombinant Human Protein, hIgG1-Fc Tag

Catalog number: 11300H02H25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Sema Domain, Seven Thrombospondin Repeats (TM) and Short Cytoplasmic Domain, (Semaphorin) 5A (SEMA5A), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the amino acids (Met 1-Thr 765) of human SEMA5A (NP_003957.2) extracellular domain fused to Fc region of human IgG1 at the C-terminus.

Activity: The activity of this protein has not been tested.

Formulation: Lyophilized in PBS, 100 mM EDTA, 0.05 mM Tween20, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: FLJ12815, SEMAF, semF, SEMA5A
Gene ID: 9037
Gene Symbol: SEMA5A
Molecular Weight: 110.7 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Other Proteins
Protein Form: Recombinant, Truncated
Protein Name: sema domain, 7 thrombospondin repeats, TM and short cytoplasmic domain, (semaphorin) 5A
Protein Subtype: Other Proteins
Protein Tag or Fusion: IgG1 Fc Tag
Purification: PAGE
Purity: ≥ 85% by SDS-PAGE
Research Category: Neurobiology, Signal Transduction
Species: Human
Tag Position: C-Terminal
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of protein in suspension buffer. Store at -20 °C..

Documents

Manuals & protocols