Product Image
Invitrogen™

Platinum Supermix (Brasil)

Platinum PCR SuperMix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification. All you have to do is add template and primers, reducing set-up time by half.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
11306161100 Reactions
11306152500 Reactions
Catalog number 11306161
Price (MXN)
2,677.95
Each
No. of Reactions:
100 Reactions
Price (MXN)
2,677.95
Each
Platinum PCR SuperMix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification with only template and primers needing to be added. The ready-to-use format reduces setup time by as much as half (see figure).

Platinum PCR SuperMix is for high-specificity DNA amplification. High specificity is provided by a mixture of recombinant Taq DNA Polymerase and Platinum monoclonal antibodies to Taq DNA Polymerase. Automatic hot start improves the specificity of PCR. Taq DNA Polymerase is activated during the initial denaturation step.

Features

  • Allows for room temperature reaction assembly
  • Amplifies fragments up to 5 kb
  • Produces products with 3'-A ends
  • Improves PCR specificity
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)1X
FormatTube
Hot StartBuilt-In Hot Start
No. of Reactions100 Reactions
Overhang3'-A
PolymeraseTaq DNA Polymerase
Product TypePlatinum Supermix
Quantity100 reactions
Reaction FormatSuperMix or Master Mix
Shipping ConditionDry Ice
Size (Final Product)5 kb or less
Starting MaterialDNA
Concentration1.1X
For Use With (Application)Hot-start PCR
GC-Rich PCR PerformanceLow
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
• Platinum PCR Supermix, 4 x 1.25 mL

Store in a non-frost-free freezer at -20°C.

Frequently asked questions (FAQs)

What is the difference between Platinum technology and AccuPrime technology?

With Platinum technology, anti-DNA polymerase antibodies bind to the enzyme until the denaturing step at 94 degrees C, when the antibodies degrade. The polymerase is now active and primer extension can occur. AccuPrime Taq combines Platinum Taq (Taq + Platinum antibodies) with proprietary thermostable AccuPrime accessory proteins. The 10X reaction buffer contains the accessory proteins which enhance specific primer-template hybridization during each cycle of PCR.