2,6-Dichloroaniline, 98%
2,6-Dichloroaniline, 98%
2,6-Dichloroaniline, 98%
Thermo Scientific Chemicals

2,6-Dichloroaniline, 98%

CAS: 608-31-1 | C6H5Cl2N | 162.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 113105000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS608-31-1
IUPAC Name2,6-dichloroaniline
Molecular FormulaC6H5Cl2N
InChI KeyJDMFXJULNGEPOI-UHFFFAOYSA-N
SMILESNC1=C(Cl)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Melting point35°C to 39°C
GC>=97.5 %
Appearance (Form)Crystalline solid
AppearanceMay darken on storage
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only