3,4-Dichloroaniline, 98%
3,4-Dichloroaniline, 98%
3,4-Dichloroaniline, 98%
Thermo Scientific Chemicals

3,4-Dichloroaniline, 98%

CAS: 95-76-1 | C6H5Cl2N | 162.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 113112500
Price (EUR)
46,40
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
46,40
Each
Chemical Identifiers
CAS95-76-1
IUPAC Name3,4-dichloroaniline
Molecular FormulaC6H5Cl2N
InChI KeySDYWXFYBZPNOFX-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to brown or grey
Appearance (Color)Containing some darker particles
Appearance (Form)Powder or crystals or flakes or fused mass
Appearance (Form)And/or chunks
AppearanceDarker particles are coarser chunks
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only