3,4-Dichlorobenzylamine, 98%
3,4-Dichlorobenzylamine, 98%
3,4-Dichlorobenzylamine, 98%
Thermo Scientific Chemicals

3,4-Dichlorobenzylamine, 98%

CAS: 102-49-8 | C7H7Cl2N | 176.05 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1132400505 mLGlass bottle
Catalog number 113240050
Price (USD)
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS102-49-8
IUPAC Name1-(3,4-dichlorophenyl)methanamine
Molecular FormulaC7H7Cl2N
InChI KeyIXHNFOOSLAWRBQ-UHFFFAOYSA-N
SMILESNCC1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.5775 to 1.5795 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only