OLIGO™ R3 Column;, 10 mm x 100mm, 7.9 ml

Catalog number: 1133246

Applied Biosystems™  Related applications: Chromatography | DNA Extraction | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific™ Oligo™ R3 bulk resin is a rigid polymeric bead designed for reversed-phase chromatography of polynucleotides and other biomolecules. The combination of 30-μm particle size, resistance to high pH, and high-phase ratio, makes Oligo R3 resin particularly suited to preparative purification of synthetic oligonucleotides and peptides. The packing consists of cross-linked poly-divinylbenzene particles. Oligo R3 bulk resin differs somewhat in selectivity from conventional silica-based, reversed-phase resins and is similar to high-carbon-loading through C18 supports. In addition, the complete lack of residual silanol activity may modify the selectivity of ionic species.

Features of Oligo R3 resin and pre-packed columns include:
• High performance chromatography resin for analytical to process scale separations
• High resolution preparative purification at high flow rates
• Chemical stability—allows aggressive cleaning and sanitization
• Poly-divinylbenzene particles—yield a robust, easily packable matrix
• Combines low back pressure, high pH stability, and high capacity

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Column Type: Analytical Column
Format: Prepacked Column
Immobilized Phase: R3
Product Line: OLIGO™
Product Size: 1 column
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols