Dichloromethane, 99.8+%, for analysis, stabilized with amylene
Dichloromethane, 99.8+%, for analysis, stabilized with amylene
Dichloromethane, 99.8+%, for analysis, stabilized with amylene
Thermo Scientific Chemicals

Dichloromethane, 99.8+%, for analysis, stabilized with amylene

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 113460010
Price (TWD)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.02 % (Coulometric)
Residue after evaporation=<0.001 %
Acidity/alkalinity=<0.0003 meq/g
Substance darkened by H2SO4=<5 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only