This product will be discontinued by May 1, 2016. We currently do not offer a direct replacement. For additional assistance, please contact our customer care team.
AFLP® Analysis System for Microorganisms
Catalog number: 11352010
Invitrogen™  Related applications: Genotyping & Genomic Profiling
Error loading your content!
  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
AFLP® Analysis Systems enable the identification of DNA polymorphisms using selective primers, classical fingerprinting techniques, and PCR to amplify a subset of genomic restriction. Three different AFLP® Analysis Systems are available:
• The AFLP® Analysis System I is designed for use with plants having genomes ranging in size from 5 x 108 to 6 x 109 bp, including tomato, corn, soybean, cucumber, lettuce, barley, cotton, oilseed rape, potato, sunflower, pepper, and brassica.
• The AFLP® Analysis System II is designed for use with plants having genomes ranging in size from 1 x 108 to 5 x 108 bp, including Arabidopsis, rice, peach, and cucumber.
• The AFLP® Analysis System for Microorganisms is designed for use with bacteria such as Agrobacterium, Xanthamonas, E. coli, and Pseudomonas, and fungi such as Collectotrichum, Pyricularia, and yeast.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Technique: AFLP
Product Size: 1 set
Genotyping Target: Microbial Genomes
Contents & storage
The AFLP® Core Reagent Kit contains reagents for template preparation, including restriction enzymes, adapters, ligase and appropriate buffers. The AFLP® Primer Kit contains primers and other reagents required for AFLP® amplification reactions. The AFLP® Analysis System I contains the AFLP® Core Reagent Kit, the AFLP® Starter Primer Kit, and a manual. The AFLP® Analysis System II contains the AFLP® Core Reagent Kit, the AFLP® Small Genome Primer Kit, and a manual. The AFLP® Analysis System for Microorganisms contains the AFLP® Core Reagent Kit, the AFLP® Microorganism Primer Kit, and a manual. Store all AFLP® Analysis Systems at -20°C. Guaranteed stable for 6 months when properly stored.
Documents
Manuals & protocols
Recommended products