Sodium Pyruvate (100 mM), 100 mL - FAQs
View additional product information for Sodium Pyruvate (100 mM) - FAQs (11360039)
1 product FAQs found
For how long is Sodium Pyruvate (100 mM) stable at 2-8 degrees C after opening the bottle?
When stored properly at 2-8 degrees C, the product is stable for 12 months from date of shipment
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.