2,3-Dichloropyridine, 99%
2,3-Dichloropyridine, 99%
2,3-Dichloropyridine, 99%
Thermo Scientific Chemicals

2,3-Dichloropyridine, 99%

CAS: 2402-77-9 | C5H3Cl2N | 147.99 g/mol
数量:
10 g
50 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 113720500
価格(JPY)
-
数量:
50 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2402-77-9
IUPAC Name2,3-dichloropyridine
Molecular FormulaC5H3Cl2N
InChI KeyMAKFMOSBBNKPMS-UHFFFAOYSA-N
SMILESClC1=CC=CN=C1Cl
さらに表示
Melting point67°C to 70°C
Appearance (Color)White to light beige
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only