2,6-Dichloropyridine, 98%
2,6-Dichloropyridine, 98%
2,6-Dichloropyridine, 98%
Thermo Scientific Chemicals

2,6-Dichloropyridine, 98%

CAS: 2402-78-0 | C5H3Cl2N | 147.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
Glass bottle
Catalog number 113731000
Price (USD)
57.90
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
57.90
Each
Chemical Identifiers
CAS2402-78-0
IUPAC Name2,6-dichloropyridine
Molecular FormulaC5H3Cl2N
InChI KeyFILKGCRCWDMBKA-UHFFFAOYSA-N
SMILESClC1=CC=CC(Cl)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Crystalline powder
Appearance (Color)White to pink to light grey
Melting point84°C to 89°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only