Diethyl Acetamidomalonate, 99+%
Diethyl Acetamidomalonate, 99+%
Diethyl Acetamidomalonate, 99+%
Thermo Scientific Chemicals

Diethyl Acetamidomalonate, 99+%

CAS: 1068-90-2 | C9H15NO5 | 217.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 113931000
Price (USD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1068-90-2
IUPAC Name1,3-diethyl 2-acetamidopropanedioate
Molecular FormulaC9H15NO5
InChI KeyISOLMABRZPQKOV-UHFFFAOYSA-N
SMILESCCOC(=O)C(NC(C)=O)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point94°C to 97°C
Appearance (Color)White to light yellow
Infrared spectrumConforms
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only