SP Scienceware™ Bench-Top Biohazard Bag Holder
Fisher Scientific™
SP Scienceware™ Bench-Top Biohazard Bag Holder
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use
Have Questions?
Catalog number 11394310
also known as FIS-11394310
Price (KRW)
-
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.
  • Rectangular steel wire frame to hold one polyethylene bag of dimensions 8.5 x 11 in. (22 x 28cm)
  • Available with epoxy coat in orange color
  • Comes with snug fitting polypropylene plastic cover to hold bag securely in place
  • Reusable and allows cleaning by hand or in automated dish washers
  • Autoclavable
  • Also used with Clavies Autoclavable Bags H13182-0812
Specifications
Product TypeBiohazard Bag Holder Kit
AutoclavableAutoclavable
ColorPink
DisposableSingle-use
For Use With (Application)Perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use
For Use With (Equipment)01-828A, 01-828-031, 01-830A, 01-826-1, 01-827A, 01-829A, 01-815-1 22 x 28cm biohazard bag, Clavies™ autoclavable bags (01-815-6)
IncludesComplete Kit: Bag Holder, Cover and Bags (100pk)
Unit SizeEach