Diethyl allylmalonate, 97%
Diethyl allylmalonate, 97%
Diethyl allylmalonate, 97%
Thermo Scientific Chemicals

Diethyl allylmalonate, 97%

CAS: 2049-80-1 | C10H16O4 | 200.23 g/mol
数量:
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 113965000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2049-80-1
IUPAC Name1,3-diethyl 2-(prop-2-en-1-yl)propanedioate
Molecular FormulaC10H16O4
InChI KeyGDWAYKGILJJNBB-UHFFFAOYSA-N
SMILESCCOC(=O)C(CC=C)C(=O)OCC
さらに表示
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4300 to 1.4320 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only