Diethyl benzylphosphonate, 99%
Diethyl benzylphosphonate, 99%
Diethyl benzylphosphonate, 99%
Thermo Scientific Chemicals

Diethyl benzylphosphonate, 99%

CAS: 1080-32-6 | C11H17O3P | 228.23 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 114171000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1080-32-6
IUPAC Namediethyl benzylphosphonate
Molecular FormulaC11H17O3P
InChI KeyAIPRAPZUGUTQKX-UHFFFAOYSA-N
SMILESCCOP(=O)(CC1=CC=CC=C1)OCC
さらに表示
Appearance (Form)Liquid
Refractive index1.4950 to 1.4980 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only