Diethylcarbamyl Chloride 99%, Thermo Scientific Chemicals
Diethylcarbamyl Chloride 99%, Thermo Scientific Chemicals
Diethylcarbamyl Chloride 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethylcarbamyl Chloride 99%, Thermo Scientific Chemicals

Catalog NumberQuantityPackaging
1142000505 mLGlass Bottle
Catalog number 114200050
Price (JPY)
-
Quantity:
5 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS88-10-8
IUPAC NameN,N-diethylcarbamoyl chloride
Molecular FormulaC5H10ClNO
InChI KeyOFCCYDUUBNUJIB-UHFFFAOYSA-N
SMILESCCN(CC)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow to brown
Refractive index1.4495 to 1.4525 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only