Carbonato de dietilo, 99 %, Thermo Scientific Chemicals
Carbonato de dietilo, 99 %, Thermo Scientific Chemicals
Carbonato de dietilo, 99 %, Thermo Scientific Chemicals
Carbonato de dietilo, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Carbonato de dietilo, 99 %, Thermo Scientific Chemicals

CAS: 105-58-8 | C5H10O3 | 118.13 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
100 mL
500 mL
1 L
2.5 L
Envase:
Frasco de vidrio
Número de catálogo 114225000
Precio (USD)
-
Cantidad:
500 mL
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS105-58-8
IUPAC Namediethyl carbonate
Molecular FormulaC5H10O3
InChI KeyOIFBSDVPJOWBCH-UHFFFAOYSA-N
SMILESCCOC(=O)OCC
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
Refractive index1.3830 to 1.3860 (20°C, 589 nm)

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Reactivo de alquilación
RUO – Research Use Only