Diethyl chloromalonate, 90%
Diethyl chloromalonate, 90%
Diethyl chloromalonate, 90%
Thermo Scientific Chemicals

Diethyl chloromalonate, 90%

CAS: 14064-10-9 | C7H11ClO4 | 194.61 g/mol
製品番号(カタログ番号) 114230050
価格(JPY)
-
数量:
5 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS14064-10-9
IUPAC Name1,3-diethyl 2-chloropropanedioate
Molecular FormulaC7H11ClO4
InChI KeyWLWCQKMQYZFTDR-UHFFFAOYSA-N
SMILESCCOC(=O)C(Cl)C(=O)OCC
さらに表示
Appearance (Color)Clear colorless to pale yellow
Infrared spectrumConforms
GC>=88.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only