Diethyl chloromalonate, 90%
Diethyl chloromalonate, 90%
Diethyl chloromalonate, 90%
Thermo Scientific Chemicals

Diethyl chloromalonate, 90%

CAS: 14064-10-9 | C7H11ClO4 | 194.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Catalog number 114230250
Price (EUR)
256,00
Each
Quantity:
25 mL
Request bulk or custom format
Price (EUR)
256,00
Each
Chemical Identifiers
CAS14064-10-9
IUPAC Name1,3-diethyl 2-chloropropanedioate
Molecular FormulaC7H11ClO4
InChI KeyWLWCQKMQYZFTDR-UHFFFAOYSA-N
SMILESCCOC(=O)C(Cl)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Infrared spectrumConforms
GC>=88.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only