Diethyl chlorophosphite, 90%, tech., Thermo Scientific Chemicals
Diethyl chlorophosphite, 90%, tech., Thermo Scientific Chemicals
Diethyl chlorophosphite, 90%, tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl chlorophosphite, 90%, tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 114251000
Price (EUR)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS589-57-1
IUPAC Namediethyl chlorophosphonite
Molecular FormulaC4H10ClO2P
InChI KeyTXHWYSOQHNMOOU-UHFFFAOYSA-N
SMILESCCOP(Cl)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Titration Argentometric>=88 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4340 to 1.4410 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only