Diethyl cyanomethylphosphonate, 96%
Diethyl cyanomethylphosphonate, 96%
Diethyl cyanomethylphosphonate, 96%
Thermo Scientific Chemicals

Diethyl cyanomethylphosphonate, 96%

CAS: 2537-48-6 | C6H12NO3P | 177.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 114270500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2537-48-6
IUPAC Namediethyl (cyanomethyl)phosphonate
Molecular FormulaC6H12NO3P
InChI KeyKWMBADTWRIGGGG-UHFFFAOYSA-N
SMILESCCOP(=O)(CC#N)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.4312 to 1.4332 (20°C, 589 nm)
Infrared spectrumConforms
GC>=95 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only