Diethylenetriamine, 98+%
Diethylenetriamine, 98+%
Diethylenetriamine, 98+%
Thermo Scientific Chemicals

Diethylenetriamine, 98+%

Diethylenetriamine, 98+%, C4H13N3, CAS Number-111-40-0, 1,4,7-triazaheptane, 2,2'-diaminodiethylamine, diethylenetriamine, 2,2'-iminodiethylamine, bis 2-aminoethyl amine, barsamide 115, ancamine deta, n,n-bis 2-aminoethyl amine, epicure t, diethylene triamine, 2.5L, 98% min. (GC), 04,255 | CAS: 111-40-0 | C4H13N3 | 103.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
10 L
Packaging:
View all
Glass bottle
Catalog number 114312500
Price (USD)
32.65
Special offer
Online exclusive
Ends: 28-Dec-2026
38.70
Save 6.05
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
32.65
Special offer
Online exclusive
Ends: 28-Dec-2026
38.70
Save 6.05
Each
Chemical Identifiers
CAS111-40-0
IUPAC Namebis(2-aminoethyl)amine
Molecular FormulaC4H13N3
InChI KeyRPNUMPOLZDHAAY-UHFFFAOYSA-N
SMILESNCCNCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<50 APHA
Infrared spectrumConforms
Refractive index1.4820 to 1.4850 (20°C, 589 nm)
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only