Diethyl fumarate, 98%
Diethyl fumarate, 98%
Diethyl fumarate, 98%
Thermo Scientific Chemicals

Diethyl fumarate, 98%

CAS: 623-91-6 | C8H12O4 | 172.18 g/mol
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 114382500
Price (JPY)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS623-91-6
IUPAC Name1,4-diethyl (2E)-but-2-enedioate
Molecular FormulaC8H12O4
InChI KeyIEPRKVQEAMIZSS-AATRIKPKSA-N
SMILESCCOC(=O)\C=C\C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4396 to 1.4456 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only