Diethyl ketomalonate, 95%
Diethyl ketomalonate, 95%
Diethyl ketomalonate, 95%
Thermo Scientific Chemicals

Diethyl ketomalonate, 95%

CAS: 609-09-6 | C7H10O5 | 174.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1144300505 gGlass bottle
Catalog number 114430050
Price (USD)
42.65
Special offer
Online exclusive
Ends: 28-Dec-2026
50.00
Save 7.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
42.65
Special offer
Online exclusive
Ends: 28-Dec-2026
50.00
Save 7.35
Each
Chemical Identifiers
CAS609-09-6
IUPAC Name1,3-diethyl 2-oxopropanedioate
Molecular FormulaC7H10O5
InChI KeyDBKKFIIYQGGHJO-UHFFFAOYSA-N
SMILESCCOC(=O)C(=O)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=94 %
Refractive index1.4150 to 1.4190 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only