N,N-Diethylnicotinamide, 97%
N,N-Diethylnicotinamide, 97%
N,N-Diethylnicotinamide, 97%
Thermo Scientific Chemicals

N,N-Diethylnicotinamide, 97%

CAS: 59-26-7 | C10H14N2O | 178.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 114470250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS59-26-7
IUPAC NameN,N-diethylpyridine-3-carboxamide
Molecular FormulaC10H14N2O
InChI KeyNCYVXEGFNDZQCU-UHFFFAOYSA-N
SMILESCCN(CC)C(=O)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Oily liquid or crystalline mass
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.5240 to 1.5260 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only