Diethyl phosphite, 98%, Thermo Scientific Chemicals
Diethyl phosphite, 98%, Thermo Scientific Chemicals
Diethyl phosphite, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl phosphite, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 114500025
Price (EUR)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS762-04-9
IUPAC Namediethoxy(oxo)-λ⁵-phosphanylium
Molecular FormulaC4H10O3P
InChI KeyLXCYSACZTOKNNS-UHFFFAOYSA-N
SMILESCCO[P+](=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4070 to 1.4090 (20°C, 589 nm)
Appearance (Color)Colorless
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only