1,3-Difluorobenzene, 99+%
1,3-Difluorobenzene, 99+%
1,3-Difluorobenzene, 99+%
Acros Organics

1,3-Difluorobenzene, 99+%

CAS: 372-18-9 | C6H4F2 | 114.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
10 mL
25 mL
Packaging:
Glass bottle
Catalog number 114601000
also known as 11460
HSN 29039990
Price (INR)
65,628.00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
65,628.00
Each
Chemical Identifiers
CAS372-18-9
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4370 to 1.4390 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only