Search
View additional product information for 11476017 - FAQs (11476017)
1 product FAQs found
The cDNA insert is cloned in the Not I/Sal I sites of the pCMVSport vectors. The Sal I site is the 5' end of the sense strand. Therefore, in order to make an in vitro transcribed/translated product from the sense strand, one would have to use the SP6 promoter.