1,8-Dihydroxyanthraquinone, 95%
1,8-Dihydroxyanthraquinone, 95%
1,8-Dihydroxyanthraquinone, 95%
Thermo Scientific Chemicals

1,8-Dihydroxyanthraquinone, 95%

CAS: 117-10-2 | C14H8O4 | 240.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 114830050
Price (EUR)
30,40
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
30,40
Each
Chemical Identifiers
CAS117-10-2
IUPAC Name1,8-dihydroxy-9,10-dihydroanthracene-9,10-dione
Molecular FormulaC14H8O4
InChI KeyQBPFLULOKWLNNW-UHFFFAOYSA-N
SMILESOC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point190°C to 195°C
HPLC>=94 %
Appearance (Color)Yellow-green to orange to brown
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only