2,5-Dihydroxybenzaldehyde, 99%
2,5-Dihydroxybenzaldehyde, 99%
2,5-Dihydroxybenzaldehyde, 99%
Thermo Scientific Chemicals

2,5-Dihydroxybenzaldehyde, 99%

CAS: 1194-98-5 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 114840050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1194-98-5
IUPAC Name2,5-dihydroxybenzaldehyde
Molecular FormulaC7H6O3
InChI KeyCLFRCXCBWIQVRN-UHFFFAOYSA-N
SMILESOC1=CC=C(O)C(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point98°C to 102°C
Appearance (Color)Yellow to khaki
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only