3,5-Dihydroxybenzoic acid, 97%
3,5-Dihydroxybenzoic acid, 97%
3,5-Dihydroxybenzoic acid, 97%
Thermo Scientific Chemicals

3,5-Dihydroxybenzoic acid, 97%

CAS: 99-10-5 | C7H6O4 | 154.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 114901000
Price (USD)
53.65
Online exclusive
58.90
Save 5.25
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
53.65
Online exclusive
58.90
Save 5.25
Each
Chemical Identifiers
CAS99-10-5
IUPAC Name3,5-dihydroxybenzoic acid
Molecular FormulaC7H6O4
InChI KeyUYEMGAFJOZZIFP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(O)=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to light pink
Infrared spectrumConforms
Melting point235°C to 242°C
Titration with NaOH96.0 to 104.0 %
HPLC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only