3,5-Dihydroxybenzoic acid, 97%
3,5-Dihydroxybenzoic acid, 97%
3,5-Dihydroxybenzoic acid, 97%
3,5-Dihydroxybenzoic acid, 97%
Thermo Scientific Chemicals

3,5-Dihydroxybenzoic acid, 97%

CAS: 99-10-5 | C7H6O4 | 154.12 g/mol
製品番号(カタログ番号) 114905000
価格(JPY)
-
見積もりを依頼する
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS99-10-5
IUPAC Name3,5-dihydroxybenzoic acid
Molecular FormulaC7H6O4
InChI KeyUYEMGAFJOZZIFP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(O)=CC(O)=C1
さらに表示
Appearance (Color)White to beige to light pink
Infrared spectrumConforms
Melting point235°C to 242°C
Titration with NaOH96.0 to 104.0 %
HPLC>=96.0 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only