4,4'-Dihydroxybenzophenone, 97%
4,4'-Dihydroxybenzophenone, 97%
4,4'-Dihydroxybenzophenone, 97%
Thermo Scientific Chemicals

4,4'-Dihydroxybenzophenone, 97%

CAS: 611-99-4 | C13H10O3 | 214.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 114920250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS611-99-4
IUPAC Name4-(4-hydroxybenzoyl)phenol
Molecular FormulaC13H10O3
InChI KeyRXNYJUSEXLAVNQ-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Appearance (Color)Off-white to beige
GC>=96.0 %
Melting point217°C to 222°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only