4,6-Dihydroxy-2-mercaptopyrimidine, 98%
4,6-Dihydroxy-2-mercaptopyrimidine, 98%
4,6-Dihydroxy-2-mercaptopyrimidine, 98%
Thermo Scientific Chemicals

4,6-Dihydroxy-2-mercaptopyrimidine, 98%

CAS: 504-17-6 | C4H4N2O2S | 144.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 114965000
Price (EUR)
533,00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
533,00
Each
Chemical Identifiers
CAS504-17-6
IUPAC Name2-sulfanylidene-1,3-diazinane-4,6-dione
Molecular FormulaC4H4N2O2S
InChI KeyRVBUGGBMJDPOST-UHFFFAOYSA-N
SMILESO=C1CC(=O)NC(=S)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
SolubilityColorless to yellow to light brown solution
Titration with NaOH97.5 to 102.5 %
Solubility(5% in 1M NaOH) Clear to slightly hazy
Appearance (Form)Powder or crystalline solid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only