Sf21 cells in Sf-900™ II SFM
Gibco™

Sf21 cells in Sf-900™ II SFM

Gibco™ Sf21 cells are commonly used to isolate and propagate recombinant baculoviral stocks and to produce recombinant proteins. Gibco™ Sf21Read more
Have Questions?
Catalog NumberQuantity
114970131.5 mL
Catalog number 11497013
Price (USD)
805.65
Online exclusive
876.00
Save 70.35
Each
Quantity:
1.5 mL
Price (USD)
805.65
Online exclusive
876.00
Save 70.35
Each
Gibco™ Sf21 cells are commonly used to isolate and propagate recombinant baculoviral stocks and to produce recombinant proteins. Gibco™ Sf21 cells are adapted to serum-free suspension culture in Gibco™ Sf-900™ II SFM, which saves significant time and expense associated with the adaptation of cultures. Gibco™ Sf21 cells (frozen in Gibco™ Sf-900™ II SFM) feature:
• Recombinant protein expression from a variety of expression systems
• Good growth in adherent or suspension culture
• Quality and performance testing

Recombinant protein expression from a variety of expression systems
High levels of protein expression in Sf21 cells can be obtained using either the BaculoDirect™ Baculovirus Expression System, the Bac-to-Bac™ Baculovirus Expression System, or the InsectDirect™ Expression System.

Good growth in adherent or suspension culture
Each vial containing 1 × 107 cells can be thawed directly into suspension or adherent culture. Protocols for adherent and suspension growth in Gibco™ Sf-900™ II SFM are available in the product manual. Cultures can easily be transferred between the two conditions for ease of workflow but we recommend the stock cultures are maintained in suspension.

Quality and performance testing
Each lot of Gibco™ Sf21 cells is tested for cell growth and viability post-recovery from cryopreservation.

Product Use
For Research Use Only. Not for any animal or human therapeutic or diagnostic use. Caution: Handle as potentially biohazardous material under at least Biosafety Level 2 containment. This product contains Dimethyl Sulfoxide (DMSO), a hazardous material. Review the Material Safety Data Sheet before handling.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Media RecommendationSf-900 II SFM (Serum-Free Media)
Product TypeInsect Cells
Quantity1.5 mL
Shipping ConditionDry Ice
Cell LineSf21
Cell TypeInsect Cells
SpeciesS. frugiperda
Unit SizeEach
Contents & Storage
Storage conditions: Liquid nitrogen (vapor phase)
Shipping conditions: Frozen

Frequently asked questions (FAQs)

What is the procedure to thaw frozen insect cells?

The following protocol describes a general procedure for thawing cryopreserved cells. For detailed protocols, always refer to the cell-specific product insert.

1. Remove the cryovial containing the frozen cells from liquid nitrogen storage and immediately place it into a 37°C water bath.
2. Quickly thaw the cells (< 1 minute) by gently swirling the vial in the 37°C water bath until there is just a small bit of ice left in the vial.
3. Transfer the vial into a laminar flow hood. Before opening, wipe the outside of the vial with 70% ethanol.
4. Transfer the desired amount of pre-warmed complete growth medium appropriate for your cell line dropwise into the centrifuge tube containing the thawed cells.
5. Centrifuge the cell suspension at approximately 200 x g for 5-10 minutes. The actual centrifugation speed and duration varies depending on the cell type.
6. After the centrifugation, check the clarity of supernatant and visibility of a complete pellet. Aseptically decant the supernatant without disturbing the cell pellet.
7. Gently resuspend the cells in complete growth medium, and transfer them into the appropriate culture vessel and into the recommended culture environment.

Note: The appropriate flask size depends on the number of cells frozen in the cryovial, and the culture environment varies based on the cell and media type.

Why does the Insect cell line manual state: "Cells should be maintained at 27 degrees C in a non-humidified environment."

Insect cells do not require CO2 or high humidity to grow, they can grow in a lab drawer at room temperature. We recommend this so people don't waste CO2 and other resources necessary for maintaining a tissue culture incubator. It should be noted, however, that the cells will grow in a humidified incubator.

What antimicrobials can be used in insect culture and at what concentration?

Many antibiotics are suitable for use with insect cells. The following antibiotics are commonly used:
- Penicillin/Streptomycine: 50-100 U/mL; 50-100 µg/mL
- Amphotericin B (Fungizone antimycotic): 0.25 µg/mL
- Gentamicin: 0.5 mL of 10 mg/mL solution in 500 mL media (final concentration: 10 µg/mL)

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is it necessary to heat-inactivate my serum before adding it to my medium?

Heat inactivation is not necessary. Our team has routinely used serum that has not been heat-inactivated, and we have not observed any effect on cell growth or morphology.

Many cells do not require heat-inactivated FBS. Some cells prefer heat-inactivated FBS. For instance, we use heat-inactivated FBS for our insect cell lines, i.e., Sf9 and Sf21 cells.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do you offer serum-free insect media?

Yes, we offer a variety of serum-free insect media. Please go here (http://www.thermofisher.com/us/en/home/life-science/cell-culture/insect-cell-culture/insect-cell-culture-misc/serum-free-media.html?icid=cvc-insect-media-c2t2) to view the media we offer and the differences between them.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (1)

Citations & References
Abstract
Conversion of non-allosteric pyruvate kinase isozyme into an allosteric enzyme by a single amino acid substitution.
Authors:Ikeda Y, Tanaka T, Noguchi T
Journal:J Biol Chem
PubMed ID:9252361
'Pyruvate kinase M1, a non-allosteric isozyme, was converted into an allosteric enzyme by replacement of an amino acid in the intersubunit contact. The substitution of Ala-398 with Arg resulted in the pronounced allosteric enzyme. The Hill coefficient and the substrate concentration giving one-half of Vmax for the mutant with respect ... More