7-(2,3-Dihydroxypropyl)theophylline, 99%
7-(2,3-Dihydroxypropyl)theophylline, 99%
7-(2,3-Dihydroxypropyl)theophylline, 99%
Thermo Scientific Chemicals

7-(2,3-Dihydroxypropyl)theophylline, 99%

CAS: 479-18-5 | C10H14N4O4 | 254.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 115051000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS479-18-5
IUPAC Name7-(2,3-dihydroxypropyl)-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular FormulaC10H14N4O4
InChI KeyKSCFJBIXMNOVSH-UHFFFAOYNA-N
SMILESCN1C2=C(N(CC(O)CO)C=N2)C(=O)N(C)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point160°C to 165°C
Titration with HClO4>=98.5 % (On dried substance)
Loss on drying=<0.5 % (1 g, 105°C)
Appearance (Color)White
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only