STD5000 and STD6000 Current-to-Pressure (I/P) Transducers
Thermo Scientific™

STD5000 and STD6000 Current-to-Pressure (I/P) Transducers

Ensure reliable signal conversion in the most demanding industrial environments with Thermo Scientific™ STD5000 and STD6000 Current-to-Pressure (I/P) Transducers.
Have Questions?
Catalog NumberEnvironmental Rating
115052IP65 (NEMA 4X)
Catalog number 115052
Price (MXN)
-
Environmental Rating:
IP65 (NEMA 4X)
This product has been discontinued however we continue to provide services and support for this product.
This product has been discontinued however we continue to provide services and support for this product.
Specifications
Environmental RatingIP65 (NEMA 4X)
Air Consumption0.04 SCFM steady state average (0.06 SCFM maximum)
Certifications/ComplianceCE marked, ATEX, CSA/US
ConnectionsSupply port; 0.25 in. NPTF (1X); Pneumatic Output Port: 0.25 in. NPTF (2X); Electrical: 0.5 in. NPTF conduit (2X); terminals 12 to 22 AWG wire
Deadband0.02% of span
DescriptionSTD5000 and STD6000 Current-to-Pressure (I/P) Transducers
Enclosure MaterialCast Aluminum with Powder-coated Epoxy
Failure ModeMechanically direct (i.e., if the input current drops below 3.7mA DC, the output will drop to 1 to 2psig regardless of direct or reverse mode selection)
Frequency Response-3 dB at 5 Hz (per ISA-S26.4.3.1 Configuration A)
Loop Load3.8 VDC + 5 ohm (19 5ohm load at 20 mA)
Operating ModesField selectable direct, reverse and/or split range; see notes in model number description
OutputsStandard: 4.0 SCFM (supply and exhaust characteristics are balanced to within ±10%); High Pressure: 4 to 8 SCFM possible (dependent on air supply and tubing size)
Repeatability0.05% of span
Stability0.5% of span/6 months
Vibration Effect<0.25% from 1 to 200Hz/1g
Weight (English)2.5 lb.
Weight (Metric)1.13 kg
Unit SizeEach