4,4'-Diiodobiphenyl, 99%
4,4'-Diiodobiphenyl, 99%
4,4'-Diiodobiphenyl, 99%
Thermo Scientific Chemicals

4,4'-Diiodobiphenyl, 99%

CAS: 3001-15-8 | C12H8I2 | 406 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 115121000
Price (USD)
290.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
290.00
Each
Chemical Identifiers
CAS3001-15-8
IUPAC Name4,4'-diiodo-1,1'-biphenyl
Molecular FormulaC12H8I2
InChI KeyGPYDMVZCPRONLW-UHFFFAOYSA-N
SMILESIC1=CC=C(C=C1)C1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=98.5 %
Appearance (Color)Light yellow to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only