N,N-Diisopropylethylamine, 99+%
N,N-Diisopropylethylamine, 99+%
N,N-Diisopropylethylamine, 99+%
Thermo Scientific Chemicals

N,N-Diisopropylethylamine, 99+%

CAS: 7087-68-5 | C8H19N | 129.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 115221000
Price (USD)
83.65
Online exclusive
92.80
Save 9.15
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
83.65
Online exclusive
92.80
Save 9.15
Each
Chemical Identifiers
CAS7087-68-5
IUPAC Nameethylbis(propan-2-yl)amine
Molecular FormulaC8H19N
InChI KeyJGFZNNIVVJXRND-UHFFFAOYSA-N
SMILESCCN(C(C)C)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4120 to 1.4150 (20°C, 589 nm)
Diisopropylamine=<0.30 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only