2',5'-Dimetoxiacetofenona, 99 %, Thermo Scientific Chemicals
2',5'-Dimetoxiacetofenona, 99 %, Thermo Scientific Chemicals
2',5'-Dimetoxiacetofenona, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2',5'-Dimetoxiacetofenona, 99 %, Thermo Scientific Chemicals

CAS: 1201-38-3 | C10H12O3 | 180.20 g/mol
Have Questions?
Número de catálogoCantidadEnvase
11534025025 gFrasco de vidrio
Número de catálogo 115340250
Precio (USD)
-
Cantidad:
25 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS1201-38-3
IUPAC Name1-(2,5-dimethoxyphenyl)ethan-1-one
Molecular FormulaC10H12O3
InChI KeyFAXUIYJKGGUCBO-UHFFFAOYSA-N
SMILESCOC1=CC=C(OC)C(=C1)C(C)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Clear yellow to yellow-green
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.5421 to 1.5441 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only