2,3-Dimethoxybenzaldehyde, 97%
2,3-Dimethoxybenzaldehyde, 97%
2,3-Dimethoxybenzaldehyde, 97%
Thermo Scientific Chemicals

2,3-Dimethoxybenzaldehyde, 97%

CAS: 86-51-1 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 115370500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS86-51-1
IUPAC Name2,3-dimethoxybenzaldehyde
Molecular FormulaC9H10O3
InChI KeyJIVGSHFYXPRRSZ-UHFFFAOYSA-N
SMILESCOC1=CC=CC(C=O)=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
Melting point48°C to 53°C
Appearance (Color)Off-white to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only