3,4-Dimethoxybenzyl alcohol, 96%
3,4-Dimethoxybenzyl alcohol, 96%
3,4-Dimethoxybenzyl alcohol, 96%
Thermo Scientific Chemicals

3,4-Dimethoxybenzyl alcohol, 96%

CAS: 93-03-8 | C9H12O3 | 168.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 115495000
Price (USD)
413.65
Special offer
Online exclusive
Ends: 28-Dec-2026
487.00
Save 73.35
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
413.65
Special offer
Online exclusive
Ends: 28-Dec-2026
487.00
Save 73.35
Each
Chemical Identifiers
CAS93-03-8
IUPAC Name(3,4-dimethoxyphenyl)methanol
Molecular FormulaC9H12O3
InChI KeyOEGPRYNGFWGMMV-UHFFFAOYSA-N
SMILESCOC1=CC=C(CO)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to golden
Appearance (Form)Viscous liquid
Infrared spectrumConforms
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only