3,4-Dimethoxycinnamic acid, 99%, predominantly trans isomer
3,4-Dimethoxycinnamic acid, 99%, predominantly trans isomer
3,4-Dimethoxycinnamic acid, 99%, predominantly trans isomer
Thermo Scientific Chemicals

3,4-Dimethoxycinnamic acid, 99%, predominantly trans isomer

CAS: 2316-26-9 | C11H12O4 | 208.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 115510050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2316-26-9
IUPAC Name(2E)-3-(3,4-dimethoxyphenyl)prop-2-enoic acid
Molecular FormulaC11H12O4
InChI KeyHJBWJAPEBGSQPR-GQCTYLIASA-N
SMILESCOC1=CC=C(\C=C\C(O)=O)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point181°C to 184°C
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH>=98.5 %
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only